Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010312.4 | NP_034442.1 | 5 | 413 | ||
GenBank mRNA | AK002604.1 | - | 5 | 408 | ||
AK146941.1 | - | 5 | 437 | |||
AK151628.1 | - | 5 | 403 | |||
AK179615.1 | - | 3 | 179 | |||
BC029077.1 | - | 4 | 253 | 4019292 | ||
BC059942.1 | - | 5 | 439 | 6529685 | ||
BC062178.1 | - | 5 | 396 | 6330944 | ||
U34960.1 | - | 4 | 233 |