Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010313.2 | NP_034443.1 | 4, 5 | 485 | ||
NM_138719.5 | NP_619733.1 | 2, 3 | 461 | |||
GenBank mRNA | AK018747.1 | - | 15 | |||
AK050564.1 | - | 2, 3 | 366 | |||
AK149908.1 | - | 2, 3 | 278 | |||
AK169323.1 | - | 2, 3 | 394 | |||
BC016135.1 | - | 4, 5 | 419 | 4506201 | ||
CK621143.1 | - | 4, 5 | 485 | |||
L34290.1 | - | 2, 3 | 287 | |||
U69145.1 | - | 3, 4 | 383 |