Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010324.2 | NP_034454.2 | 2 | 326 | ||
GenBank mRNA | AK146445.1 | - | 2 | 228 | ||
AK147807.1 | - | 2 | 247 | |||
AK147934.1 | - | 2 | 231 | |||
BC002057.1 | - | 2 | 214 | 3489628 | ||
CK334037.1 | - | 2 | 326 | |||
J02623.1 | - | 2 | 230 |