Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145914.2 | NP_666019.1 | 2 | 239 | ||
XM_005250568.4 | XP_005250625.1 | 3 | 1381 | |||
XM_006716113.3 | XP_006716176.1 | 2 | 1220 | |||
XM_017012584.1 | XP_016868073.1 | 3 | 1345 | |||
XM_017012585.1 | XP_016868074.1 | 2 | 263 | |||
GenBank mRNA | AK075324.1 | - | 3 | 339 | ||
AK292267.1 | - | 2 | 259 | |||
AK301871.1 | - | 3 | 365 | |||
AK313884.1 | - | 2 | 259 | |||
AL136865.1 | - | 1 | 192 | |||
BC047309.1 | - | 2 | 232 | 5111890 | ||
BI764964.1 | - | 2 | 239 | |||
CR749301.1 | - | 2 | 234 |