Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010361.2 | NP_034491.2 | 2 | 744 | ||
GenBank mRNA | AK002392.1 | - | 2 | 274 | ||
AK079739.1 | - | 2 | 326 | |||
AK158939.1 | - | 735 | ||||
BC012707.1 | - | 2 | 302 | 3994154 | ||
U48420.1 | - | 2 | 280 | |||
X98056.1 | - | 2 | 149 |