Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010370.2 | NP_034500.1 | 1, 2 | 108 | ||
GenBank mRNA | AK146132.1 | - | 1, 2 | 105 | ||
AK156728.1 | - | 1, 2 | 107 | |||
BC061146.1 | - | 1, 2 | 108 | 30296854 | ||
M26183.1 | - | 1, 2 | 96 | |||
X14799.1 | - | 1, 2 | 110 |