Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010372.2 | NP_034502.2 | 2 | 219 | ||
GenBank mRNA | AK131671.1 | - | 2 | 220 | ||
BC141122.1 | - | 2 | 174 | 9055734 | ||
BC145433.1 | - | 174 | 9053991 | |||
BC145434.1 | - | 2 | 174 | |||
J03255.1 | - | 2 | 234 |