Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010411.2 | NP_034541.2 | 12 | 1034 | ||
GenBank mRNA | AF074881.1 | - | 12 | 981 | ||
AF074882.1 | - | 6 | 408 | |||
AF098295.1 | - | 12 | 959 | |||
AF125536.1 | - | 12 | 966 | |||
AK037478.1 | - | 509 | ||||
AK145158.1 | - | 12 | 1000 | |||
AK206656.1 | - | 5 | 321 | |||
AK208847.1 | - | 1 | 47 | |||
BC139300.1 | - | 12 | 1062 | 8862322 | ||
BC139301.1 | - | 12 | 1062 | 8862323 | ||
JN651901.1 | - | 7 | 433 | |||
JN651902.1 | - | 6 | 391 | |||
JN651903.1 | - | 6 | 391 |