Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010287.2 | NP_034417.1 | 2, 3 | 801 | ||
GenBank mRNA | AK035356.1 | - | 2, 3 | 846 | ||
AK081276.1 | - | 2, 3 | 814 | |||
M80481.1 | - | 2, 3 | 802 |
Set Membership: |
G-protein coupled receptor |