Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010299.3 | NP_034429.1 | 3, 4 | 614 | ||
GenBank mRNA | AK149642.1 | - | 3, 4 | 507 | ||
AK152256.1 | - | 3, 4 | 506 | |||
AK210428.1 | - | 1, 2 | 191 | |||
BC004651.1 | - | 3, 4 | 482 | 3482848 | ||
CT010289.1 | - | 3, 4 | 435 | |||
L19526.1 | - | 3, 4 | 448 | |||
U09816.1 | - | 3, 4 | 491 |