Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010312.4 | NP_034442.1 | 9 | 965 | ||
GenBank mRNA | AK002604.1 | - | 9 | 961 | ||
AK146941.1 | - | 9 | 989 | |||
AK151628.1 | - | 9 | 955 | |||
BC029077.1 | - | 8 | 805 | 4019292 | ||
BC059942.1 | - | 9 | 991 | 6529685 | ||
BC062178.1 | - | 9 | 948 | 6330944 | ||
U34960.1 | - | 8 | 785 | |||
U38505.1 | - | 2 | 242 |