Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010322.3 | NP_034452.3 | 2 | 335 | ||
GenBank mRNA | AF110769.1 | - | 2 | 299 | ||
AJ132012.1 | - | 2 | 130 | |||
AK004630.1 | - | 2 | 309 | |||
AK004771.1 | - | 2 | 315 | |||
AK010896.1 | - | 2 | 309 | |||
AK150810.1 | - | 2 | 293 | |||
AK166997.1 | - | 2 | 307 | |||
AK183472.1 | - | 2 | 125 | |||
BC003225.1 | - | 2 | 307 | |||
BC025972.1 | - | 2 | 292 | 4020505 | ||
BI990299.1 | - | 2 | 335 | |||
BY775400.1 | - | 2 | 331 |