Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010330.4 | NP_034460.3 | 4 | 734 | ||
GenBank mRNA | AK079570.1 | - | 4 | 676 | ||
AK088480.1 | - | 4 | 562 | |||
AK138271.1 | - | 4 | 683 | |||
AK148449.1 | - | 4 | 562 | |||
AK148540.1 | - | 4 | 562 | |||
AK150625.1 | - | 4 | 563 | |||
AK161679.1 | - | 4 | 562 | |||
AK166602.1 | - | 4 | 562 | |||
AK167997.1 | - | 4 | 594 | |||
AK200349.1 | - | 2 | 231 | |||
BC014858.1 | - | 4 | 553 | 4500922 | ||
J03535.2 | - | 4 | 561 |