Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080552.1 | NP_001074021.1 | 2 | 278 | ||
NM_001080553.1 | NP_001074022.1 | 2 | 431 | |||
NM_010352.2 | NP_034482.2 | 3 | 491 | |||
GenBank mRNA | BC023009.1 | - | 2 | 363 | 5364798 | |
D87325.1 | - | 3 | 370 |