Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077353.2 | NP_001070821.1 | 8 | 810 | ||
NM_001288617.1 | NP_001275546.1 | 8 | 855 | |||
NM_010356.4 | NP_034486.2 | 7 | 715 | |||
GenBank mRNA | AK002478.1 | - | 6 | 663 | ||
AK014076.1 | - | 7 | 688 | |||
AK149543.1 | - | 7 | 680 | |||
BC147272.1 | - | 7 | 678 | 9056327 | ||
BC147273.1 | - | 7 | 678 | 9056339 | ||
M73483.1 | - | 6 | 646 | |||
X65021.1 | - | 6 | 631 |