Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF054823.1 | - | 3, 4 | 375 | ||
AK079456.1 | - | 3, 4 | 404 | |||
AK088296.1 | - | 3, 4 | 404 | |||
AK144680.1 | - | 3, 4 | 406 | |||
AK146112.1 | - | 3, 4 | 405 | |||
AK149320.1 | - | 3, 4 | 406 | |||
AK185133.1 | - | 1, 2 | 31 | |||
BC012638.1 | - | 2, 3 | 264 | 4191809 |