Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gusb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010368.1 | NP_034498.1 | 3 | 470 | ||
GenBank mRNA | AK076109.1 | - | 3 | 502 | ||
AK136519.1 | - | 3 | 527 | |||
AK146247.1 | - | 3 | 692 | |||
AK149814.1 | - | 3 | 500 | |||
AK150048.1 | - | 3 | 471 | |||
AK151802.1 | - | 3 | 471 | |||
AK152206.1 | - | 3 | 471 | |||
AK159163.1 | - | 3 | 471 | |||
AK159526.1 | - | 3 | 471 | |||
AK159564.1 | - | 3 | 462 | |||
AK159804.1 | - | 3 | 471 | |||
AK162436.1 | - | 3 | 471 | |||
BC004616.1 | - | 3 | 507 | |||
BC018202.1 | - | 3 | 607 | |||
BC071226.1 | - | 3 | 491 | 5325429 | ||
J03047.1 | - | 3 | 470 | |||
M19279.1 | - | 3 | 470 | |||
M28540.1 | - | 3 | 470 | |||
M28541.1 | - | 3 | 470 | |||
M63836.1 | - | 3 | 470 |