Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010411.2 | NP_034541.2 | 12, 13 | 1046 | ||
GenBank mRNA | AF074881.1 | - | 12, 13 | 993 | ||
AF074882.1 | - | 6, 7 | 420 | |||
AF098295.1 | - | 12, 13 | 971 | |||
AF125536.1 | - | 12, 13 | 978 | |||
AK145158.1 | - | 12, 13 | 1012 | |||
AK206656.1 | - | 5, 6 | 333 | |||
AK208847.1 | - | 1, 2 | 59 | |||
BC139300.1 | - | 12, 13 | 1074 | 8862322 | ||
BC139301.1 | - | 12, 13 | 1074 | 8862323 | ||
JN651903.1 | - | 6, 7 | 403 |