Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077696.1 | NP_001071164.1 | 6 | 909 | ||
NM_001284248.1 | NP_001271177.1 | 8 | 1421 | |||
NM_001284249.1 | NP_001271178.1 | 7 | 1018 | |||
NM_001284250.1 | NP_001271179.1 | 6 | 906 | |||
NM_010412.3 | NP_034542.3 | 6 | 906 | |||
GenBank mRNA | AF006602.2 | - | 5 | 622 | ||
AF207748.1 | - | 5 | 541 | |||
AK145972.1 | - | 6 | 872 | |||
AK146483.1 | - | 6 | 845 | |||
AK155335.1 | - | 6 | 880 | |||
AK156008.1 | - | 3 | 1157 | |||
AK157159.1 | - | 3 | 1157 | |||
BC049926.1 | - | 6 | 842 | |||
BC058554.1 | - | 8 | 1373 | |||
BC060609.1 | - | 5 | 700 | 5720438 |