Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010287.2 | NP_034417.1 | 3 | 843 | ||
GenBank mRNA | AK030359.1 | - | 1 | 1559 | ||
AK035356.1 | - | 3 | 888 | |||
AK041527.1 | - | 2 | 733 | |||
AK081276.1 | - | 3 | 856 | |||
M80481.1 | - | 3 | 844 |
Set Membership: |
G-protein coupled receptor |