Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023560.3 | NP_001018854.2 | 3, 4 | 815 | ||
NM_001111039.2 | NP_001104509.1 | 3, 4 | 812 | |||
NM_001287421.1 | NP_001274350.1 | 3, 4 | 301 | |||
NM_152736.5 | NP_689949.3 | 3, 4 | 410 | |||
XM_011514862.2 | XP_011513164.1 | 3, 4 | 1014 | |||
XM_011514864.1 | XP_011513166.1 | 4, 5 | 591 | |||
XM_017011264.1 | XP_016866753.1 | 3, 4 | 1012 | |||
GenBank mRNA | AK095948.1 | - | 3, 4 | 362 | ||
AK293524.1 | - | 3, 4 | 535 | |||
AK295393.1 | - | 2, 3 | 965 | |||
AK301081.1 | - | 3, 4 | 706 | |||
AK315645.1 | - | 3, 4 | 274 | |||
AL526328.3 | - | 3, 4 | 667 | |||
AL832741.1 | - | 2, 3 | 1003 | |||
BC013951.2 | - | 3, 4 | 266 | 2963683 | ||
BC013962.2 | - | 3, 4 | 266 | 2963683 | ||
BX537535.1 | - | 3, 4 | 683 | |||
DA167287.1 | - | 3, 4 | 299 | |||
DA341149.1 | - | 3, 4 | 299 | |||
DA566660.1 | - | 3, 4 | 411 | |||
DB262147.1 | - | 3, 4 | 277 | |||
Z11773.1 | - | 2, 3 | 332 |