Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030718.1 | - | 1 | 1851 | ||
AK047197.1 | - | 8 | 2158 | |||
AK051968.1 | - | 1 | 203 |