Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010313.2 | NP_034443.1 | 11 | 1073 | ||
NM_138719.5 | NP_619733.1 | 9 | 1049 | |||
GenBank mRNA | AK018747.1 | - | 7 | 603 | ||
AK050564.1 | - | 9 | 954 | |||
AK148175.1 | - | 9 | 944 | |||
AK149908.1 | - | 9 | 866 | |||
AK169323.1 | - | 9 | 982 | |||
BC016135.1 | - | 11 | 1007 | 4506201 | ||
L34290.1 | - | 9 | 875 | |||
U69145.1 | - | 10 | 971 |