Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038655.1 | NP_001033744.1 | 4 | 2754 | ||
NM_010319.3 | NP_034449.2 | 4 | 2691 | |||
GenBank mRNA | AK141541.1 | - | 4 | 2663 | ||
AK147843.1 | - | 4 | 2655 | |||
AK148026.1 | - | 1 | 2430 | |||
AK158963.1 | - | 4 | 2754 | |||
BC034108.1 | - | 3 | 2618 | 4503666 |