Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010324.2 | NP_034454.2 | 8 | 1122 | ||
GenBank mRNA | AK146445.1 | - | 8 | 1024 | ||
AK147807.1 | - | 8 | 1045 | |||
AK147934.1 | - | 8 | 1027 | |||
BC002057.1 | - | 8 | 1010 | 3489628 | ||
J02623.1 | - | 8 | 1026 |