Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010330.4 | NP_034460.3 | 9 | 1464 | ||
GenBank mRNA | AK079570.1 | - | 9 | 1406 | ||
AK088480.1 | - | 9 | 1292 | |||
AK138271.1 | - | 9 | 1413 | |||
AK148449.1 | - | 9 | 1292 | |||
AK148540.1 | - | 9 | 1292 | |||
AK150625.1 | - | 9 | 1293 | |||
AK161679.1 | - | 9 | 1292 | |||
AK166602.1 | - | 9 | 1292 | |||
BC014858.1 | - | 9 | 1283 | 4500922 | ||
J03535.2 | - | 9 | 1291 |