Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111268.1 | NP_001104738.1 | 9 | 1538 | ||
NM_010349.2 | NP_034479.2 | 9 | 1538 | |||
GenBank mRNA | BC131639.1 | - | 8 | 1219 | ||
BC131640.1 | - | 8 | 1219 | |||
BC148311.2 | - | 8 | 1219 | |||
D10054.1 | - | 9 | 1451 | |||
X66117.1 | - | 8 | 1275 |