Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gusb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010368.1 | NP_034498.1 | 12 | 2145 | ||
GenBank mRNA | AK041058.1 | - | 11 | 1964 | ||
AK136519.1 | - | 12 | 2202 | |||
AK146247.1 | - | 12 | 2366 | |||
AK150048.1 | - | 12 | 2146 | |||
AK151802.1 | - | 12 | 2147 | |||
AK152206.1 | - | 12 | 2147 | |||
AK159163.1 | - | 12 | 2146 | |||
AK159526.1 | - | 12 | 2146 | |||
AK159564.1 | - | 12 | 2135 | |||
AK159804.1 | - | 12 | 2146 | |||
AK162436.1 | - | 12 | 2146 | |||
AK208599.1 | - | 1 | 146 | |||
BC071226.1 | - | 12 | 2166 | 5325429 | ||
J03047.1 | - | 12 | 2145 | |||
M19279.1 | - | 12 | 2145 | |||
M28540.1 | - | 12 | 2145 | |||
M28541.1 | - | 12 | 2145 | |||
M63836.1 | - | 12 | 2145 |