Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168271.1 | NP_001161743.1 | 2, 3 | 429 | ||
NM_153002.2 | NP_694547.2 | 2, 3 | 429 | |||
XM_005247165.2 | XP_005247222.1 | 3, 4 | 706 | |||
XM_017005795.1 | XP_016861284.1 | 3, 4 | 742 | |||
XM_017005796.1 | XP_016861285.1 | 3, 4 | 751 | |||
XM_017005797.1 | XP_016861286.1 | 3, 4 | 706 | |||
GenBank mRNA | AF488739.1 | - | 2, 3 | 232 | ||
BC113701.1 | - | 2, 3 | 252 | 8322753 | ||
BC143606.1 | - | 2, 3 | 252 | 9052119 |