Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h1t | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010377.3 | NP_034507.2 | 1 | 429 | ||
GenBank mRNA | BC119305.1 | - | 1 | 399 | 8734054 | |
BC119307.1 | - | 1 | 399 | 8734056 | ||
BV210386.1 | - | 1 | 263 |