Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-DMb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154923.1 | - | 6 | 919 | ||
BC052864.1 | - | 7 | 1070 | |||
BC141105.1 | - | 6 | 881 | 9055717 | ||
BC145385.1 | - | 6 | 881 | 9053943 | ||
BC145386.1 | - | 6 | 881 | 9053944 | ||
BC171960.1 | - | 6 | 881 | 9054246 |
Gene Symbol: H2-DMb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010387.3 | NP_034517.2 | 6 | 1023 | ||
GenBank mRNA | AK032745.1 | - | 6 | 1024 | ||
AK169969.1 | - | 6 | 1004 | |||
AK170866.1 | - | 6 | 1006 | |||
BC002237.1 | - | 985 | 3491702 | |||
BC003718.1 | - | 6 | 963 | 3486844 | ||
X62743.1 | - | 923 |