Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010421.4 | NP_034551.2 | 9 | 1091 | ||
GenBank mRNA | AK075895.1 | - | 9 | 1067 | ||
AK075911.1 | - | 9 | 1092 | |||
AK080777.1 | - | 9 | 1064 | |||
AK144168.1 | - | 9 | 1219 | |||
AK150444.1 | - | 9 | 1062 | |||
AK159091.1 | - | 10 | 1062 | |||
AK159814.1 | - | 9 | 1077 | |||
AK159890.1 | - | 9 | 1064 | |||
AK160112.1 | - | 9 | 1051 | |||
AK167013.1 | - | 9 | 1077 | |||
AK168186.1 | - | 9 | 1091 | |||
AK170903.1 | - | 9 | 1078 | |||
AK213356.1 | - | 1 | 46 | |||
BC010755.1 | - | 9 | 1060 | 4225233 | ||
U07631.1 | - | 9 | 1083 | |||
X64331.1 | - | 9 | 1034 |