Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136065.2 | NP_001129537.1 | 2 | 567 | ||
NM_001294143.1 | NP_001281072.1 | 2 | 643 | |||
NM_001294144.1 | NP_001281073.1 | 2 | 643 | |||
NM_010433.2 | NP_034563.2 | 2 | 643 | |||
GenBank mRNA | AF077659.1 | - | 2 | 406 | ||
AF170301.1 | - | 2 | 393 | |||
AF170302.1 | - | 2 | 393 | |||
AF208292.1 | - | 2 | 480 | |||
AF273680.1 | - | 2 | 364 | |||
AF333791.1 | - | 2 | 270 | |||
AF333792.1 | - | 2 | 270 | |||
AK016742.1 | - | 2 | 570 | |||
BC031904.1 | - | 2 | 376 | 5368577 |