Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010449.4 | NP_034579.3 | 1 | 452 | ||
GenBank mRNA | AK083575.1 | - | 1 | 453 | ||
BC138097.1 | - | 1 | 386 | 8861116 | ||
BC138098.1 | - | 1 | 389 | 8861117 | ||
M22115.1 | - | 1 | 421 |