Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ids | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010498.3 | NP_034628.2 | 1, 2 | 284 | ||
GenBank mRNA | AF538870.1 | - | 1, 2 | 283 | ||
AK085237.1 | - | 2, 3 | 161 | |||
AK132287.1 | - | 2, 3 | 164 | |||
AK166178.1 | - | 1, 2 | 162 | |||
AK166443.1 | - | 2, 3 | 113 | |||
L07921.1 | - | 1, 2 | 161 |