Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010504.2 | NP_034634.1 | 1 | 174 | ||
GenBank mRNA | BC116217.2 | - | 1 | 129 | 40091191 | |
BC119349.1 | - | 1 | 174 | 8734098 | ||
BC119351.1 | - | 1 | 174 | 8734100 |