Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010559.2 | NP_034689.2 | 2 | 281 | ||
GenBank mRNA | AK046337.1 | - | 2 | 280 | ||
AK082270.1 | - | 2 | 281 | |||
BC019388.1 | - | 2 | 275 | |||
X51975.1 | - | 2 | 172 | |||
X51976.1 | - | 161 | ||||
X53802.1 | - | 2 | 281 |