Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136066.2 | NP_001129538.1 | 3 | 266 | ||
NM_010443.2 | NP_034573.2 | 3 | 282 | |||
GenBank mRNA | AF029874.1 | - | 3 | 279 | ||
AF054669.1 | - | 3 | 283 | |||
AF054670.1 | - | 3 | 217 | |||
AK032175.1 | - | 3 | 224 | |||
AK134536.1 | - | 3 | 241 | |||
AK141426.1 | - | 3 | 260 | |||
AK152936.1 | - | 3 | 205 | |||
BC002011.1 | - | 3 | 238 | 3484259 | ||
BC055742.1 | - | 1 | 1413 | |||
CA465845.1 | - | 3 | 297 |