Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039129.4 | NP_001034218.1 | 3 | 359 | ||
NM_010447.5 | NP_034577.1 | 3 | 359 | |||
GenBank mRNA | AK007802.1 | - | 3 | 305 | ||
AK088308.1 | - | 3 | 304 | |||
AK131999.1 | - | 3 | 325 | |||
AK135391.1 | - | 3 | 303 | |||
AK136113.1 | - | 3 | 327 | |||
AK146126.1 | - | 3 | 303 | |||
AK146630.1 | - | 3 | 325 | |||
AK152684.1 | - | 3 | 303 | |||
AK161730.1 | - | 3 | 303 | |||
AK166901.1 | - | 3 | 303 | |||
AK166989.1 | - | 3 | 301 | |||
AK167161.1 | - | 3 | 303 | |||
AK167810.1 | - | 3 | 303 | |||
AK167811.1 | - | 3 | 303 | |||
AK167820.1 | - | 3 | 305 | |||
AK167913.1 | - | 3 | 303 | |||
AK168420.1 | - | 3 | 303 | |||
AK168531.1 | - | 3 | 306 | |||
AK168733.1 | - | 3 | 303 | |||
AK169216.1 | - | 3 | 303 | |||
AK187359.1 | - | 1 | 29 | |||
AK187715.1 | - | 1 | 37 | |||
BC052723.1 | - | 3 | 289 | |||
BC080675.1 | - | 3 | 285 | 6308746 | ||
BC083136.1 | - | 3 | 270 | 6511966 | ||
BC089340.1 | - | 3 | 278 | 6402927 | ||
BC092395.1 | - | 3 | 279 | 6403376 | ||
BY067931.1 | - | 3 | 361 | |||
D86728.1 | - | 3 | 266 | |||
D86729.1 | - | 3 | 266 | |||
M99167.1 | - | 3 | 301 |