Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048061.1 | NP_001041526.1 | 3 | 546 | ||
NM_010448.3 | NP_034578.1 | 3 | 546 | |||
GenBank mRNA | AK013384.1 | - | 3 | 513 | ||
AK013709.1 | - | 512 | ||||
AK076132.1 | - | 3 | 509 | |||
AK165609.1 | - | 4 | 493 | |||
BC043069.1 | - | 3 | 500 | 6402930 | ||
CA325334.1 | - | 2 | 244 | |||
D90151.1 | - | 3 | 501 | |||
DQ397821.1 | - | 2 | 308 | |||
DQ397822.1 | - | 2 | 308 | |||
L36663.1 | - | 505 |