Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010453.5 | NP_034583.1 | 2 | 1123 | ||
GenBank mRNA | AK144356.1 | - | 2 | 864 | ||
BC011063.1 | - | 2 | 947 | 3985274 | ||
M28021.1 | - | 2 | 1123 | |||
M36604.1 | - | 2 | 1000 | |||
X16840.1 | - | 2 | 1000 |