Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010461.2 | NP_034591.1 | 2 | 1675 | ||
GenBank mRNA | AB221615.1 | - | 2 | 614 | ||
AK131612.1 | - | 2 | 550 | |||
BC139209.1 | - | 2 | 855 | 8862231 | ||
BC139212.1 | - | 3 | 795 | 8862234 | ||
X13721.1 | - | 2 | 1674 |
Gene Symbol: Hoxd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290730.1 | NP_001277659.1 | 2 | 820 | ||
NM_001290731.1 | NP_001277660.1 | 3 | 451 | |||
NM_008276.3 | NP_032302.2 | 2 | 823 | |||
GenBank mRNA | AK016033.1 | - | 2 | 552 | ||
BX513411.1 | - | 3 | 475 |