Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010466.2 | NP_034596.1 | 1 | 628 | ||
GenBank mRNA | AB221617.1 | - | 1 | 391 | ||
AK051173.1 | - | 1 | 84 | |||
AK079948.1 | - | 1 | 217 | |||
AK080107.1 | - | 1 | 190 | |||
X07439.1 | - | 1 | 628 | |||
X07646.1 | - | 1 | 856 |