Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010475.1 | NP_034605.1 | 4 | 532 | ||
GenBank mRNA | AK019406.1 | - | 1 | 50 | ||
AK143346.1 | - | 4 | 481 | |||
BC125657.1 | - | 4 | 531 | 40130172 | ||
BC125659.1 | - | 4 | 531 | 40130174 | ||
X89627.1 | - | 4 | 532 |