Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038698.1 | NP_001033787.1 | 2 | 466 | ||
NM_001163397.1 | NP_001156869.1 | 2 | 302 | |||
NM_001163399.1 | NP_001156871.1 | 2 | 552 | |||
NM_010488.4 | NP_034618.2 | 2 | 552 | |||
GenBank mRNA | AF041341.1 | - | 2 | 314 | ||
AK014133.1 | - | 2 | 320 | |||
AK079088.1 | - | 2 | 457 | |||
BB648788.1 | - | 2 | 553 | |||
BC048159.1 | - | 2 | 525 | 5703005 | ||
BC051987.1 | - | 223 | ||||
BC052451.1 | - | 2 | 429 | 6409658 | ||
CF726962.1 | - | 2 | 302 | |||
CN663544.1 | - | 2 | 466 | |||
D31953.1 | - | 2 | 299 |