Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142386.2 | NP_001135858.1 | 3, 4 | 546 | ||
NM_005391.4 | NP_005382.1 | 3, 4 | 546 | |||
GenBank mRNA | AK301965.1 | - | 3, 4 | 549 | ||
BC015948.1 | - | 3, 4 | 493 | 3922929 | ||
BG717206.1 | - | 3, 4 | 557 | |||
L42452.1 | - | 3, 4 | 339 |