Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033343.3 | NP_203129.1 | 3 | 552 | ||
XM_011510105.2 | XP_011508407.1 | 3 | 786 | |||
XM_011510106.2 | XP_011508408.1 | 3 | 409 | |||
XM_011510108.2 | XP_011508410.1 | 3 | 192 | |||
XM_017002755.1 | XP_016858244.1 | 2 | 904 | |||
GenBank mRNA | AB055703.1 | - | 2 | 255 | ||
AB055704.1 | - | 2 | 268 | |||
AF179849.1 | - | 3 | 556 | |||
AF282899.1 | - | 2 | 251 | |||
AY053457.1 | - | 3 | 344 | |||
BC011759.2 | - | 3 | 363 | 3529089 | ||
KU178818.1 | - | 3 | 320 | |||
KU178819.1 | - | 4 | 371 |