Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010554.4 | NP_034684.2 | 5, 6 | 563 | ||
GenBank mRNA | AK156449.1 | - | 5, 6 | 564 | ||
BC003727.1 | - | 4, 5 | 532 | 3599550 | ||
M14639.1 | - | 1, 2 | 197 | |||
X01450.1 | - | 5, 6 | 563 |