Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098203.1 | NP_001091673.1 | 2 | 2714 | ||
NM_010430.2 | NP_034560.2 | 2 | 3708 | |||
GenBank mRNA | AF036334.1 | - | 2 | 2503 |