Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291065.1 | NP_001277994.1 | 2 | 1919 | ||
NM_001291067.1 | NP_001277996.1 | 2 | 1987 | |||
NM_010446.3 | NP_034576.2 | 3 | 1977 | |||
GenBank mRNA | CK023291.1 | - | 1 | 549 | ||
L10409.1 | - | 2 | 1878 | |||
X74937.1 | - | 3 | 1977 |